Product Details

SNP ID
rs141371836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43460726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCCAGGTCGCGGAGCAAAGGTC[A/G]ATCACGTTCTCGATCAAAAGGCAGG
Phenotype
MIM: 601944
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRSF6 PubMed Links

Gene Details

Gene
SRSF6
Gene Name
serine and arginine rich splicing factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006275.5 868 Missense Mutation CAA,CGA Q233R NP_006266.2

View Full Product Details