Product Details

SNP ID
rs141543812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:24963960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTCTCCGGAAGGCACCGCTGTC[A/G]CTGAGTTCTAGGACGAGGCTGTACC
Phenotype
MIM: 615884 MIM: 603253
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APMAP PubMed Links

Gene Details

Gene
APMAP
Gene Name
adipocyte plasma membrane associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020531.2 1205 Silent Mutation AGC,AGT S368S NP_065392.1
XM_005260763.3 1205 UTR 3 XP_005260820.1
Gene
CST7
Gene Name
cystatin F
There are no transcripts associated with this gene.

View Full Product Details