Product Details

SNP ID
rs141834554
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56365252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCCTTGGGCATATTTTACATG[A/G]TTGTGTCAAGGTAAGATTTTTGACA
Phenotype
MIM: 603072
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AURKA PubMed Links

Gene Details

Gene
AURKA
Gene Name
aurora kinase A
There are no transcripts associated with this gene.

Gene
FAM210B
Gene Name
family with sequence similarity 210 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080821.2 431 Missense Mutation ATT,GTT I118V NP_543011.2

View Full Product Details