Product Details

SNP ID
rs141869916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32633175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCAAACTATATCAGTGACTCAC[C/T]ATCCCAGTTTGCCTGGGACTGTGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf203 PubMed Links

Gene Details

Gene
C20orf203
Gene Name
chromosome 20 open reading frame 203
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182584.2 3243 UTR 3 NP_872390.2

View Full Product Details