Product Details

SNP ID
rs141934538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:41361358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCGGGCAATGTCCTGCGTGTG[C/T]TGGGCCAGCGTACGATTCAGCTGGT
Phenotype
MIM: 608929 MIM: 605520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMILIN3 PubMed Links

Gene Details

Gene
EMILIN3
Gene Name
elastin microfibril interfacer 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052846.1 2435 Silent Mutation CAA,CAG Q737Q NP_443078.1
Gene
LPIN3
Gene Name
lipin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301860.1 2435 Intron NP_001288789.1
XM_005260516.2 2435 Intron XP_005260573.1
XM_006723863.2 2435 Intron XP_006723926.1
XM_011528996.1 2435 Intron XP_011527298.1
XM_011528997.2 2435 Intron XP_011527299.1
XM_011528998.2 2435 Intron XP_011527300.1
XM_011528999.2 2435 Intron XP_011527301.1
XM_011529000.1 2435 Intron XP_011527302.1
XM_011529001.1 2435 Intron XP_011527303.1
XM_011529002.2 2435 Intron XP_011527304.1
XM_011529003.1 2435 Intron XP_011527305.1
XM_011529004.1 2435 Intron XP_011527306.1
XM_011529005.2 2435 Intron XP_011527307.1
XM_011529006.2 2435 Intron XP_011527308.1
XM_017028020.1 2435 Intron XP_016883509.1

View Full Product Details