Product Details

SNP ID
rs141946874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49364550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTCCATTGGAAAACCACTTGGT[G/T]GTCCCCCCCATGGGTCGTGTGTGAG
Phenotype
MIM: 600397
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNB1 PubMed Links

Gene Details

Gene
KCNB1
Gene Name
potassium voltage-gated channel subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004975.3 11174 UTR 3 NP_004966.1
XM_006723784.3 11174 UTR 3 XP_006723847.1
XM_011528799.2 11174 UTR 3 XP_011527101.1

View Full Product Details