Product Details

SNP ID
rs142022310
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3044467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGCTGCTGCTGACTGCCCACC[C/T]TGGACCCTCAGAGGCTCAGCACTGG
Phenotype
MIM: 602352 MIM: 611988 MIM: 176884
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GNRH2 PubMed Links
Additional Information
For this assay, SNP(s) [rs6051545] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GNRH2
Gene Name
gonadotropin releasing hormone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310220.1 104 Missense Mutation CCT,CTT P18L NP_001297149.1
NM_001501.1 104 Missense Mutation CCT,CTT P18L NP_001492.1
NM_178331.1 104 Missense Mutation CCT,CTT P18L NP_847901.1
NM_178332.1 104 Missense Mutation CCT,CTT P18L NP_847902.1
XM_017027823.1 104 Missense Mutation CCT,CTT P18L XP_016883312.1
Gene
MRPS26
Gene Name
mitochondrial ribosomal protein S26
There are no transcripts associated with this gene.

Gene
PTPRA
Gene Name
protein tyrosine phosphatase, receptor type A
There are no transcripts associated with this gene.

View Full Product Details