Product Details
- SNP ID
-
rs142022479
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44337392 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACCACATCCGGGCCAGTGTGTAC[C/G]CACCTGCCGCCAACATGGAATACAT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
R3HDML
PubMed Links
Gene Details
- Gene
- R3HDML
- Gene Name
- R3H domain containing like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178491.3 |
235 |
Missense Mutation |
CCA,GCA |
P79A |
NP_848586.1 |
View Full Product Details