Product Details

SNP ID
rs142217156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46560032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTCCAGGGGGACTCAGAGGGTC[C/T]GAAATGTGTCATTGGCCAAGGTGGG
Phenotype
MIM: 606411
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OCSTAMP PubMed Links

Gene Details

Gene
OCSTAMP
Gene Name
osteoclast stimulatory transmembrane protein
There are no transcripts associated with this gene.

Gene
SLC13A3
Gene Name
solute carrier family 13 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011554.2 1666 Missense Mutation CAG,CGG Q553R NP_001011554.1
NM_001193339.1 1666 Missense Mutation CAG,CGG Q550R NP_001180268.1
NM_001193340.1 1666 Missense Mutation CAG,CGG Q518R NP_001180269.1
NM_001193342.1 1666 Missense Mutation CAG,CGG Q502R NP_001180271.1
NM_022829.5 1666 Missense Mutation CAG,CGG Q600R NP_073740.2

View Full Product Details