Product Details
- SNP ID
-
rs142360045
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:647127 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCCTACCTGGCCTAACCCCATAC[C/T]AGCAGTGCAGACAAGGAGGCACTCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SRXN1
PubMed Links
Gene Details
- Gene
- SRXN1
- Gene Name
- sulfiredoxin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080725.2 |
2186 |
UTR 3 |
|
|
NP_542763.1 |
View Full Product Details