Product Details

SNP ID
rs142616480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46178069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGCTGTCGCAGCCACAGATGC[A/G]GATGGTGAGCGTGCCTGTGCTGCTC
Phenotype
MIM: 609920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH22 PubMed Links

Gene Details

Gene
CDH22
Gene Name
cadherin 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021248.2 2394 Missense Mutation CGC,TGC R598C NP_067071.1
XM_011528994.2 2394 Missense Mutation CGC,TGC R598C XP_011527296.1
XM_011528995.1 2394 Missense Mutation CGC,TGC R477C XP_011527297.1

View Full Product Details