Product Details

SNP ID
rs142982868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62878847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCACACCGGCCTCCGGGGCGGC[A/G]GCTAAGACCTGTAAAAGCAGCTTTT
Phenotype
MIM: 604140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIDO1 PubMed Links

Gene Details

Gene
DIDO1
Gene Name
death inducer-obliterator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193369.1 7441 UTR 3 NP_001180298.1
NM_001193370.1 7441 Intron NP_001180299.1
NM_022105.4 7441 Intron NP_071388.2
NM_033081.2 7441 UTR 3 NP_149072.2
NM_080796.3 7441 Intron NP_542986.1
NM_080797.3 7441 Intron NP_542987.2
XM_011528504.1 7441 UTR 3 XP_011526806.1
XM_011528505.1 7441 UTR 3 XP_011526807.1
XM_011528506.1 7441 UTR 3 XP_011526808.1
XM_011528507.1 7441 UTR 3 XP_011526809.1
XM_011528508.1 7441 UTR 3 XP_011526810.1
XM_011528509.2 7441 Intron XP_011526811.1

View Full Product Details