Product Details
- SNP ID
-
rs143061754
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:1537194 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCGGAGGCAGCAGCAGGGTTGGA[C/T]GAAATAGTTTGTGCTGTTTCTCTCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SIRPD
PubMed Links
Gene Details
- Gene
- SIRPD
- Gene Name
- signal regulatory protein delta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178460.2 |
582 |
Missense Mutation |
ATC,GTC |
I180V |
NP_848555.2 |
View Full Product Details