Product Details

SNP ID
rs143518276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45841873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTGAGACCTGGGGCTTCACTC[A/G]GATCACGCCCTCCTGGGCACAGGTC
Phenotype
MIM: 608123 MIM: 191039
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACOT8 PubMed Links

Gene Details

Gene
ACOT8
Gene Name
acyl-CoA thioesterase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005469.3 1764 Nonsense Mutation CGA,TGA R309* NP_005460.2
XM_005260239.3 1764 Nonsense Mutation CGA,TGA R256* XP_005260296.1
XM_011528480.2 1764 Nonsense Mutation CGA,TGA R174* XP_011526782.1
Gene
SNX21
Gene Name
sorting nexin family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042632.2 1764 UTR 3 NP_001036097.1
NM_001042633.2 1764 UTR 3 NP_001036098.1
NM_033421.3 1764 UTR 3 NP_219489.1
NM_152897.2 1764 Intron NP_690857.1
XM_005260608.4 1764 UTR 3 XP_005260665.1
XM_011529095.2 1764 UTR 3 XP_011527397.1
XM_011529096.2 1764 Intron XP_011527398.1
XM_011529098.2 1764 Intron XP_011527400.1
XM_011529099.2 1764 Intron XP_011527401.1
XM_017028120.1 1764 Intron XP_016883609.1
Gene
TNNC2
Gene Name
troponin C2, fast skeletal type
There are no transcripts associated with this gene.

View Full Product Details