Product Details
- SNP ID
-
rs143595580
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45207086 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAACATCATCTTTGTACTTTCCCT[C/G]CTCCTCATCTTGGAGAAGCAAGCAG
- Phenotype
-
MIM: 182140
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SEMG1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2233882,rs2233883] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEMG1
- Gene Name
- semenogelin I
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003007.4 |
123 |
Silent Mutation |
CTC,CTG |
L11L |
NP_002998.1 |
View Full Product Details