Product Details
- SNP ID
-
rs143890191
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45175883 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTTCCTGTTAAAGGTCAAGACAC[C/T]GTCAAAGGCCGTGTTCCATTCAATG
- Phenotype
-
MIM: 182257
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PI3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2664581] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PI3
- Gene Name
- peptidase inhibitor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002638.3 |
126 |
Silent Mutation |
ACC,ACT |
T34T |
NP_002629.1 |
View Full Product Details