Product Details

SNP ID
rs144136156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34834457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTAGTTTCCTGGGGTCACTTCT[A/G]GCCATCCTGACTGTATTGTTCACTA
Phenotype
MIM: 605299
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMGB3P1 PubMed Links

Gene Details

Gene
HMGB3P1
Gene Name
high mobility group box 3 pseudogene 1
There are no transcripts associated with this gene.

Gene
NCOA6
Gene Name
nuclear receptor coactivator 6
There are no transcripts associated with this gene.

View Full Product Details