Product Details
- SNP ID
-
rs144164912
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23877615 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGAAGTAGTAGTTCACCCCACCCA[C/T]GATCTACACGCGTGGAAGAGCAGGA
- Phenotype
-
MIM: 123858
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST5
PubMed Links
Gene Details
- Gene
- CST5
- Gene Name
- cystatin D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001900.4 |
302 |
Missense Mutation |
ATG,GTG |
M79V |
NP_001891.2 |
View Full Product Details