Product Details

SNP ID
rs144281638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63742048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTTTGGGGAGCCCACCCTGAGA[A/G]AAAGGAAGGTGAGCTTGGGGGCGGC
Phenotype
MIM: 609809 MIM: 609493 MIM: 614639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIME1 PubMed Links

Gene Details

Gene
LIME1
Gene Name
Lck interacting transmembrane adaptor 1
There are no transcripts associated with this gene.

Gene
SLC2A4RG
Gene Name
SLC2A4 regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020062.3 626 Missense Mutation AAA,GAA K191E NP_064446.2
Gene
ZBTB46
Gene Name
zinc finger and BTB domain containing 46
There are no transcripts associated with this gene.

Gene
ZGPAT
Gene Name
zinc finger CCCH-type and G-patch domain containing
There are no transcripts associated with this gene.

View Full Product Details