Product Details

SNP ID
rs144288124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1535793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCCTGAGGCTCTGTTTTCTTA[C/T]CTATGTAATGTATACAATGTGGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIRPD PubMed Links

Gene Details

Gene
SIRPD
Gene Name
signal regulatory protein delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178460.2 Intron NP_848555.2

View Full Product Details