Product Details
- SNP ID
-
rs144300885
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:50587252 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTTTAAAATATTGTGATTTCAA[C/T]ATCTGCCTCCTGGCAAAAGACTTCT
- Phenotype
-
MIM: 176885
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM65C
PubMed Links
Gene Details
- Gene
- FAM65C
- Gene Name
- family with sequence similarity 65 member C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290268.1 |
3235 |
Missense Mutation |
ATT,GTT |
I945V |
NP_001277197.1 |
NM_080829.3 |
3235 |
Missense Mutation |
ATT,GTT |
I941V |
NP_543019.2 |
XM_005260294.3 |
3235 |
Missense Mutation |
ATT,GTT |
I945V |
XP_005260351.1 |
XM_006723713.3 |
3235 |
Missense Mutation |
ATT,GTT |
I940V |
XP_006723776.1 |
XM_011528578.2 |
3235 |
Missense Mutation |
ATT,GTT |
I945V |
XP_011526880.1 |
XM_011528579.2 |
3235 |
Missense Mutation |
ATT,GTT |
I941V |
XP_011526881.1 |
XM_011528580.2 |
3235 |
Missense Mutation |
ATT,GTT |
I941V |
XP_011526882.1 |
XM_011528581.2 |
3235 |
Missense Mutation |
ATT,GTT |
I941V |
XP_011526883.1 |
XM_011528584.2 |
3235 |
Intron |
|
|
XP_011526886.1 |
XM_011528585.2 |
3235 |
Missense Mutation |
ATT,GTT |
I608V |
XP_011526887.1 |
XM_011528586.2 |
3235 |
Intron |
|
|
XP_011526888.1 |
XM_017027681.1 |
3235 |
Missense Mutation |
ATT,GTT |
I715V |
XP_016883170.1 |
XM_017027682.1 |
3235 |
Intron |
|
|
XP_016883171.1 |
- Gene
- MIR645
- Gene Name
- microRNA 645
There are no transcripts associated with this gene.
- Gene
- PTPN1
- Gene Name
- protein tyrosine phosphatase, non-receptor type 1
There are no transcripts associated with this gene.
View Full Product Details