Product Details

SNP ID
rs144518160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18813881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGGACCTCTGGTGGGCCAGTT[C/T]GAGGTTCTGGGTTCAGTTCCAGAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf78 PubMed Links

Gene Details

Gene
C20orf78
Gene Name
chromosome 20 open reading frame 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242671.2 156 Intron NP_001229600.1
NM_001302818.1 156 Intron NP_001289747.1
Gene
SCP2D1
Gene Name
SCP2 sterol binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178483.2 156 Silent Mutation TTC,TTT F22F NP_848578.1

View Full Product Details