Product Details

SNP ID
rs144821472
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33410163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACACCTCCTGCACACCCTCGGC[A/G]GCCCGGTGACAGCCATCCACAAGCT
Phenotype
MIM: 608200 MIM: 601017
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CDK5RAP1 PubMed Links

Gene Details

Gene
CDK5RAP1
Gene Name
CDK5 regulatory subunit associated protein 1
There are no transcripts associated with this gene.

Gene
SNTA1
Gene Name
syntrophin alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003098.2 1442 Silent Mutation GCC,GCT A403A NP_003089.1
XM_005260517.1 1442 Silent Mutation GCC,GCT A403A XP_005260574.1
XM_011529007.1 1442 Silent Mutation GCC,GCT A403A XP_011527309.1
XM_011529008.1 1442 Silent Mutation GCC,GCT A403A XP_011527310.1

View Full Product Details