Product Details
- SNP ID
-
rs144851310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:187950 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCCTGCAGCACACTCAGCTTCTCA[C/T]CAGAATGTTGATGTGGCTTCTTAAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB128
PubMed Links
Gene Details
- Gene
- DEFB128
- Gene Name
- defensin beta 128
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037732.2 |
276 |
Missense Mutation |
GAT,GGT |
D73G |
NP_001032821.1 |
View Full Product Details