Product Details

SNP ID
rs144884181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:16273108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACTCAGGCACTAGGTATGGAAA[C/T]GTGAGGTGGCCCGGGCCCGCTGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF16B PubMed Links

Gene Details

Gene
KIF16B
Gene Name
kinesin family member 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199865.1 4130 UTR 3 NP_001186794.1
NM_001199866.1 4130 Intron NP_001186795.1
NM_024704.4 4130 UTR 3 NP_078980.3
XM_005260750.3 4130 Intron XP_005260807.1
XM_005260751.4 4130 Intron XP_005260808.1
XM_005260752.3 4130 Intron XP_005260809.1
XM_005260753.3 4130 UTR 3 XP_005260810.1
XM_005260754.3 4130 UTR 3 XP_005260811.1
XM_005260755.3 4130 UTR 3 XP_005260812.1
XM_006723588.3 4130 UTR 3 XP_006723651.1
XM_017027926.1 4130 UTR 3 XP_016883415.1

View Full Product Details