Product Details
- SNP ID
-
rs145029548
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:35463597 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGTGCTGCAGTACCGAAGCTGGT[G/T]CCAAGAGCTGGAGAAGCGGCTAGAA
- Phenotype
-
MIM: 609689
MIM: 601146
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CEP250
PubMed Links
Gene Details
- Gene
- CEP250
- Gene Name
- centrosomal protein 250
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318219.1 |
856 |
UTR 5 |
|
|
NP_001305148.1 |
NM_007186.5 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
NP_009117.2 |
XM_005260262.4 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_005260319.1 |
XM_005260263.4 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_005260320.1 |
XM_005260264.4 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_005260321.1 |
XM_006723690.3 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_006723753.1 |
XM_006723691.1 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_006723754.1 |
XM_006723692.3 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_006723755.1 |
XM_006723693.3 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_006723756.1 |
XM_006723694.3 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_006723757.1 |
XM_011528517.2 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_011526819.1 |
XM_011528518.2 |
856 |
UTR 5 |
|
|
XP_011526820.1 |
XM_011528519.2 |
856 |
Intron |
|
|
XP_011526821.1 |
XM_017027617.1 |
856 |
Missense Mutation |
TGC,TTC |
C70F |
XP_016883106.1 |
XM_017027618.1 |
856 |
Intron |
|
|
XP_016883107.1 |
XM_017027619.1 |
856 |
Intron |
|
|
XP_016883108.1 |
- Gene
- GDF5
- Gene Name
- growth differentiation factor 5
There are no transcripts associated with this gene.
- Gene
- MIR1289-1
- Gene Name
- microRNA 1289-1
There are no transcripts associated with this gene.
View Full Product Details