Product Details

SNP ID
rs145060259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44756173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGGGCCCCACGGTGCTCTCGAG[A/G]GACAACTGGGATGCCTGCCGGAGCA
Phenotype
MIM: 607368 MIM: 611601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNK15 PubMed Links

Gene Details

Gene
KCNK15
Gene Name
potassium two pore domain channel subfamily K member 15
There are no transcripts associated with this gene.

Gene
KCNK15-AS1
Gene Name
KCNK15 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
RIMS4
Gene Name
regulating synaptic membrane exocytosis 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205317.1 838 Silent Mutation TCC,TCT S258S NP_001192246.1
NM_182970.3 838 Silent Mutation TCC,TCT S257S NP_892015.1

View Full Product Details