Product Details

SNP ID
rs145151680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18794069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTGATGCGGGCTTCGTGAGGC[A/G]GAAGTGACGCAGTGTTGTCGTCTCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00652 PubMed Links

Gene Details

Gene
LINC00652
Gene Name
long intergenic non-protein coding RNA 652
There are no transcripts associated with this gene.

Gene
LOC100270804
Gene Name
uncharacterized LOC100270804
There are no transcripts associated with this gene.

View Full Product Details