Product Details

SNP ID
rs145357542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5938667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCATGGCAACGGTGAAGCCGGA[C/G]AGAAGATAACCCCCACCTCCACTCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRMT6 PubMed Links

Gene Details

Gene
TRMT6
Gene Name
tRNA methyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281467.1 1611 Silent Mutation CTC,CTG L284L NP_001268396.1
NM_015939.4 1611 Silent Mutation CTC,CTG L454L NP_057023.2
XM_017027875.1 1611 Silent Mutation CTC,CTG L272L XP_016883364.1

View Full Product Details