Product Details

SNP ID
rs145410144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33740905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCTGCACTTCTTTTTTCAGGAA[C/T]GGACAATCAGACCGTTCTGGCTGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF341 PubMed Links

Gene Details

Gene
ZNF341
Gene Name
zinc finger protein 341
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282933.1 297 Missense Mutation ACG,ATG T12M NP_001269862.1
NM_001282935.1 297 UTR 5 NP_001269864.1
NM_032819.4 297 Missense Mutation ACG,ATG T12M NP_116208.3

View Full Product Details