Product Details
- SNP ID
-
rs146087665
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:17946186 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCCTATGGATGTGGACATGGACC[A/G]GACTTGTGATCCCTGAAACCTGCAT
- Phenotype
-
MIM: 605937
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX5
PubMed Links
Gene Details
- Gene
- SNX5
- Gene Name
- sorting nexin 5
View Full Product Details