Product Details
- SNP ID
-
rs146415539
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:157559 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGCCTGGCCATGGGGCTCTTCAT[G/T]ATCATTGCAATTCTGCTGTTCCAGA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB127
PubMed Links
Gene Details
- Gene
- DEFB127
- Gene Name
- defensin beta 127
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_139074.3 |
90 |
Missense Mutation |
ATG,ATT |
M5I |
NP_620713.1 |
View Full Product Details