Product Details
- SNP ID
-
rs146538492
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23566004 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGCTCTGTGCTTTCTTGGAAATG[A/G]CAGTTGTCAATGTCGTCTTCAAATT
- Phenotype
-
MIM: 616536
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CST9L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2295564] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CST9L
- Gene Name
- cystatin 9-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080610.2 |
623 |
Silent Mutation |
TGC,TGT |
C108C |
NP_542177.1 |
View Full Product Details