Product Details

SNP ID
rs146614184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44621129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCATCTGTGTTGAGCGAGTAGTT[A/C]GCCTGGTCATTTTTGAGCCTGCAGA
Phenotype
MIM: 608958 MIM: 604932
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADA PubMed Links

Gene Details

Gene
ADA
Gene Name
adenosine deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000022.3 1015 Silent Mutation GCG,GCT A288A NP_000013.2
NM_001322050.1 1015 Silent Mutation GCG,GCT A153A NP_001308979.1
NM_001322051.1 1015 Silent Mutation GCG,GCT A264A NP_001308980.1
Gene
PKIG
Gene Name
protein kinase (cAMP-dependent, catalytic) inhibitor gamma
There are no transcripts associated with this gene.

View Full Product Details