Product Details

SNP ID
rs146821586
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45886938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATGCGGGAATTTGTGGCAGTTT[C/T]GGCTGTATTCCTTCCGTAGTGTCTC
Phenotype
MIM: 613111 MIM: 608597
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CTSA PubMed Links
Additional Information
For this assay, SNP(s) [rs3827040] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CTSA
Gene Name
cathepsin A
There are no transcripts associated with this gene.

Gene
NEURL2
Gene Name
neuralized E3 ubiquitin protein ligase 2
There are no transcripts associated with this gene.

Gene
SPATA25
Gene Name
spermatogenesis associated 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080608.3 272 Missense Mutation CAA,CGA Q88R NP_542175.1
Gene
ZSWIM1
Gene Name
zinc finger SWIM-type containing 1
There are no transcripts associated with this gene.

Gene
ZSWIM3
Gene Name
zinc finger SWIM-type containing 3
There are no transcripts associated with this gene.

View Full Product Details