Product Details

SNP ID
rs147072738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62956785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGGCCCGAGTGCCAGGCATGGA[C/T]GGGGACGCTGCTGCTGGGCACATGC
Phenotype
MIM: 611625 MIM: 612107
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GID8 PubMed Links

Gene Details

Gene
GID8
Gene Name
GID complex subunit 8 homolog
There are no transcripts associated with this gene.

Gene
SLC17A9
Gene Name
solute carrier family 17 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302643.1 326 Missense Mutation ACG,ATG T21M NP_001289572.1
NM_022082.3 326 Missense Mutation ACG,ATG T27M NP_071365.3
XM_011528978.2 326 UTR 5 XP_011527280.1

View Full Product Details