Product Details

SNP ID
rs147253525
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49636105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCCAGTGCTGACGAAGGAAGTC[A/C]CCAAGCTCACTCCCTCAGTTCCAGC
Phenotype
MIM: 604016
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
B4GALT5 PubMed Links

Gene Details

Gene
B4GALT5
Gene Name
beta-1,4-galactosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004776.3 1568 UTR 3 NP_004767.1

View Full Product Details