Product Details
- SNP ID
-
rs147311278
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:5302041 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCAATTGTGTGACACCAGTGGGT[A/C]ACTTCAGCCTGATACAGTCCACCTC
- Phenotype
-
MIM: 607123
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PROKR2
PubMed Links
Gene Details
- Gene
- PROKR2
- Gene Name
- prokineticin receptor 2
There are no transcripts associated with this gene.
View Full Product Details