Product Details
- SNP ID
-
rs147397940
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:36653649 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACTTTCTCCAGAGCCGAGGCTACT[C/T]GAGGTTGAGTGGGTTCGTGATCGGG
- Phenotype
-
MIM: 605273
MIM: 606577
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NDRG3
PubMed Links
Gene Details
- Gene
- NDRG3
- Gene Name
- NDRG family member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022477.3 |
1087 |
Silent Mutation |
TCA,TCG |
S321S |
NP_071922.2 |
NM_032013.3 |
1087 |
Silent Mutation |
TCA,TCG |
S333S |
NP_114402.1 |
XM_006723837.3 |
1087 |
Silent Mutation |
TCA,TCG |
S346S |
XP_006723900.1 |
XM_006723838.3 |
1087 |
Silent Mutation |
TCA,TCG |
S337S |
XP_006723901.1 |
XM_011528925.2 |
1087 |
Silent Mutation |
TCA,TCG |
S316S |
XP_011527227.1 |
XM_011528926.2 |
1087 |
Silent Mutation |
TCA,TCG |
S303S |
XP_011527228.1 |
XM_011528927.2 |
1087 |
Silent Mutation |
TCA,TCG |
S334S |
XP_011527229.1 |
XM_011528928.2 |
1087 |
Silent Mutation |
TCA,TCG |
S254S |
XP_011527230.1 |
XM_017027978.1 |
1087 |
Silent Mutation |
TCA,TCG |
S346S |
XP_016883467.1 |
XM_017027979.1 |
1087 |
Silent Mutation |
TCA,TCG |
S333S |
XP_016883468.1 |
XM_017027980.1 |
1087 |
Silent Mutation |
TCA,TCG |
S324S |
XP_016883469.1 |
- Gene
- SLA2
- Gene Name
- Src like adaptor 2
There are no transcripts associated with this gene.
View Full Product Details