Product Details
- SNP ID
-
rs147513716
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44337366 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACATGAATGCCTTACTGGATTATC[A/T]CAACCACATCCGGGCCAGTGTGTAC
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
R3HDML
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1884612] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- R3HDML
- Gene Name
- R3H domain containing like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178491.3 |
209 |
Missense Mutation |
CAC,CTC |
H70L |
NP_848586.1 |
View Full Product Details