Product Details

SNP ID
rs147565637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:6041286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCTCAAGCCCGCCCTGTTGGC[C/T]GCCAGCACGCACACGCGGTAGGTGG
Phenotype
MIM: 608188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRLS1 PubMed Links

Gene Details

Gene
CRLS1
Gene Name
cardiolipin synthase 1
There are no transcripts associated with this gene.

Gene
LRRN4
Gene Name
leucine rich repeat neuronal 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152611.4 1195 Silent Mutation GCA,GCG A653A NP_689824.2
XM_011529183.2 1195 Silent Mutation GCA,GCG A316A XP_011527485.1

View Full Product Details