Product Details

SNP ID
rs147794171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:42075113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTATCACTTCTAGACATGTATAC[A/G]TGGAAAACCTCCAACAGGGAAACTT
Phenotype
MIM: 608712
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPRT PubMed Links

Gene Details

Gene
PTPRT
Gene Name
protein tyrosine phosphatase, receptor type T
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007050.5 10276 UTR 3 NP_008981.4
NM_133170.3 10276 UTR 3 NP_573400.3
XM_017027611.1 10276 UTR 3 XP_016883100.1
XM_017027612.1 10276 UTR 3 XP_016883101.1
XM_017027613.1 10276 UTR 3 XP_016883102.1

View Full Product Details