Product Details

SNP ID
rs148103941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45176005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCAGTCTCCACTAAGCCTGGCT[C/T]CTGCCCCATTATCTTGATCCGGTGC
Phenotype
MIM: 182257
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PI3 PubMed Links

Gene Details

Gene
PI3
Gene Name
peptidase inhibitor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002638.3 248 Missense Mutation TCC,TTC S75F NP_002629.1

View Full Product Details