Product Details

SNP ID
rs148131170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:51599199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGAATCTCCGAGCAAAGCCAGGT[A/G]CAGGGTTTGCAAAGTCTAAACAGGA
Phenotype
MIM: 609126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP9A PubMed Links

Gene Details

Gene
ATP9A
Gene Name
ATPase phospholipid transporting 9A (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006045.2 5226 UTR 3 NP_006036.1
XM_011528481.2 5226 UTR 3 XP_011526783.1

View Full Product Details