Product Details
- SNP ID
-
rs148175822
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:38213517 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTTCTCCAGGAGCATCCGGATC[C/T]CCTCATGCCTGATCCGGGCCTCCTC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1755
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3746471] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIAA1755
- Gene Name
- KIAA1755
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001029864.1 |
3400 |
Missense Mutation |
GAG,GGG |
E1043G |
NP_001025035.1 |
View Full Product Details