Product Details

SNP ID
rs148298666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33657983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGGGTGTCCGGGGCCCGCAGG[C/T]GGTCGATGAGGATGCGCTGGAAGGC
Phenotype
MIM: 603672 MIM: 612478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTL10 PubMed Links

Gene Details

Gene
ACTL10
Gene Name
actin like 10
There are no transcripts associated with this gene.

Gene
C20orf144
Gene Name
chromosome 20 open reading frame 144
There are no transcripts associated with this gene.

Gene
CBFA2T2
Gene Name
CBFA2/RUNX1 translocation partner 2
There are no transcripts associated with this gene.

Gene
NECAB3
Gene Name
N-terminal EF-hand calcium binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031231.3 1125 Missense Mutation CAC,CGC H340R NP_112508.3
NM_031232.3 1125 Missense Mutation CAC,CGC H374R NP_112509.3
XM_005260510.1 1125 Missense Mutation CAC,CGC H322R XP_005260567.1
XM_011528991.1 1125 Missense Mutation CAC,CGC H179R XP_011527293.1
XM_011528992.2 1125 Intron XP_011527294.1
XM_017028015.1 1125 Missense Mutation CAC,CGC H179R XP_016883504.1
XM_017028016.1 1125 Missense Mutation CAC,CGC H179R XP_016883505.1

View Full Product Details