Product Details

SNP ID
rs148697745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57649397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACTACCCCATGAACTTCTGCC[A/G]TCCACTGGACATGAAGCTGAGGACA
Phenotype
MIM: 606564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PMEPA1 PubMed Links

Gene Details

Gene
PMEPA1
Gene Name
prostate transmembrane protein, androgen induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001255976.1 3913 UTR 3 NP_001242905.1
NM_020182.4 3913 UTR 3 NP_064567.2
NM_199169.2 3913 UTR 3 NP_954638.1
NM_199170.2 3913 UTR 3 NP_954639.1
NM_199171.2 3913 UTR 3 NP_954640.1

View Full Product Details