Product Details

SNP ID
rs148704271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34550028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTGGAAAAGCAGCTGTGGACCC[A/T]GGTCTGTCTGGCCCGCCTGCCGTTG
Phenotype
MIM: 607167 MIM: 601242
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DYNLRB1 PubMed Links

Gene Details

Gene
DYNLRB1
Gene Name
dynein light chain roadblock-type 1
There are no transcripts associated with this gene.

Gene
MAP1LC3A
Gene Name
microtubule associated protein 1 light chain 3 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032514.3 218 Intron NP_115903.1
NM_181509.2 218 Silent Mutation CCA,CCT P17P NP_852610.1
XM_011529083.2 218 Silent Mutation CCA,CCT P17P XP_011527385.1
XM_011529084.2 218 Silent Mutation CCA,CCT P17P XP_011527386.1
XM_011529085.2 218 Silent Mutation CCA,CCT P17P XP_011527387.1

View Full Product Details