Product Details

SNP ID
rs148863832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46178019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGAGGCGGCCATGACAAAGGC[A/C]GTGGTGTTGCAGGACTGGATGGTGC
Phenotype
MIM: 609920
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDH22 PubMed Links

Gene Details

Gene
CDH22
Gene Name
cadherin 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021248.2 2444 Silent Mutation NP_067071.1
XM_011528994.2 2444 Silent Mutation XP_011527296.1
XM_011528995.1 2444 Silent Mutation XP_011527297.1

View Full Product Details