Product Details

SNP ID
rs148905697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23491856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACAACAAAGAGAGCGAGGACAA[A/G]TATGTCTTCCTGGTGGTCAAGACAC
Phenotype
MIM: 608683
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CST8 PubMed Links

Gene Details

Gene
CST8
Gene Name
cystatin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281730.1 574 Silent Mutation AAA,AAG K63K NP_001268659.1
NM_005492.3 574 Silent Mutation AAA,AAG K63K NP_005483.1

View Full Product Details